Assay Details
| Gene Symbol: LOC100795446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255157.1 | NP_001242086.1 | 3 | 1219 | 126 | |
| GenBank mRNA | BT097869.1 | - | 3 | 1219 | 126 | |
More Information
Set Membership: |
|

LOC100795446
Genomic Map