Assay Details
| Gene Symbol: LOC100780620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255262.2 | NP_001242191.1 | 3 | 1214 | 132 | |
| GenBank mRNA | BT096700.1 | - | 3 | 1215 | 132 | |
More Information
Set Membership: |
|

LOC100780620
Genomic Map