Assay Details
| Gene Symbol: FKF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248957.1 | NP_001235886.1 | 2 | 1006 | 71 | |
| GenBank mRNA | DQ371902.1 | - | 2 | 1006 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FKF1| Gene Symbol: FKF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248957.1 | NP_001235886.1 | 2 | 1006 | 71 | |
| GenBank mRNA | DQ371902.1 | - | 2 | 1006 | 71 | |
Set Membership: |
|