Assay Details
| Gene Symbol: Eny2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850176.2 | XP_004850233.1 | 3 - 4 | 320 | 86 | |
| XM_004850177.3 | XP_004850234.1 | 3 - 4 | 308 | 86 | ||
More Information
Set Membership: |
|

Eny2
Genomic Map