Assay Details
| Gene Symbol: Eif4h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840038.3 | XP_004840095.1 | 1 - 2 | 168 | 75 | |
| XM_004840039.3 | XP_004840096.1 | 1 - 2 | 168 | 75 | ||
More Information
Set Membership: |
|

Eif4h
Genomic Map