Assay Details
| Gene Symbol: Znf532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858413.3 | XP_004858470.1 | 4 - 5 | 3330 | 62 | |
| XM_004858414.3 | XP_004858471.1 | 5 - 6 | 2932 | 62 | ||
| XM_004858415.3 | XP_004858472.1 | 4 - 5 | 2850 | 62 | ||
| XM_021239292.1 | XP_021094951.1 | 5 - 6 | 3412 | 62 | ||
| XM_021239293.1 | XP_021094952.1 | 4 - 5 | 2845 | 62 | ||
| XM_021239294.1 | XP_021094953.1 | 4 - 5 | 2871 | 62 | ||
| XM_021239296.1 | XP_021094955.1 | 4 - 5 | 3139 | 62 | ||
More Information
Set Membership: |
|

Znf532
Genomic Map