Assay Details
| Gene Symbol: Ccnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863083.3 | XP_004863140.1 | 8 - 9 | 963 | 71 | |
| XM_021243154.1 | XP_021098813.1 | 7 - 8 | 590 | 71 | ||
| XM_021243155.1 | XP_021098814.1 | 7 - 8 | 799 | 71 | ||
More Information
Set Membership: |
|

Ccnt1
Genomic Map