Assay Details
| Gene Symbol: Hoxd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013076958.2 | XP_012932412.1 | 1 - 2 | 591 | 69 | |
| XM_013076959.2 | XP_012932413.1 | 1 - 2 | 591 | 69 | ||
More Information
Set Membership: |
|

Hoxd3
Genomic Map