Assay Details
| Gene Symbol: Camlg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836228.2 | XP_004836285.1 | 1 - 2 | 452 | 102 | |
| XM_021238714.1 | XP_021094373.1 | 1 - 2 | 312 | 102 | ||
More Information
Set Membership: |
|

Camlg
Genomic Map