Assay Details
| Gene Symbol: Hbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839669.3 | XP_004839726.1 | 9 - 10 | 1590 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hbp1| Gene Symbol: Hbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839669.3 | XP_004839726.1 | 9 - 10 | 1590 | 97 | |
Set Membership: |
|