Assay Details
| Gene Symbol: Hexb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310312.1 | NP_001297241.1 | 5 - 6 | 690 | 84 | |
| GenBank mRNA | KM486720.1 | - | 5 - 6 | 690 | 84 | |
More Information
Set Membership: |
|

Hexb
Genomic Map