Assay Details
| Gene Symbol: Sh3rf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305106.1 | NP_001292035.1 | 4 - 5 | 1069 | 67 | |
| XM_021266101.1 | XP_021121760.1 | 5 - 6 | 1356 | 67 | ||
| XM_021266102.1 | XP_021121761.1 | 6 - 7 | 1459 | 67 | ||
| XM_021266103.1 | XP_021121762.1 | 5 - 6 | 1248 | 67 | ||
| XM_021266104.1 | XP_021121763.1 | 5 - 6 | 1197 | 67 | ||
| XM_021266106.1 | XP_021121765.1 | 5 - 6 | 1210 | 67 | ||
| XM_021266107.1 | XP_021121766.1 | 5 - 6 | 1198 | 67 | ||
| XM_021266108.1 | XP_021121767.1 | 4 - 5 | 1260 | 67 | ||
| XM_021266109.1 | XP_021121768.1 | 5 - 6 | 1356 | 67 | ||
| XM_021266110.1 | XP_021121769.1 | 5 - 6 | 1356 | 67 | ||
| XM_021266111.1 | XP_021121770.1 | 4 - 5 | 1260 | 67 | ||
More Information
Set Membership: |
|

Sh3rf2
Genomic Map