Assay Details
| Gene Symbol: Atp5f1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310268.1 | NP_001297197.1 | 3 - 4 | 246 | 88 | |
| XM_004854539.3 | XP_004854596.1 | 3 - 4 | 357 | 88 | ||
| XM_004854540.3 | XP_004854597.1 | 3 - 4 | 357 | 88 | ||
| GenBank mRNA | KM486686.1 | - | 3 - 4 | 246 | 88 | |
More Information
Set Membership: |
|

Atp5f1c
Genomic Map