Assay Details
| Gene Symbol: Asic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001279840.1 | NP_001266769.1 | 1 - 2 | 663 | 96 | |
| GenBank mRNA | JN703734.1 | - | 1 - 2 | 663 | 96 | |
More Information
Set Membership: |
|

Asic1
Genomic Map