Assay Details
| Gene Symbol: Rnf128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861471.2 | XP_004861528.1 | 3 - 4 | 1028 | 64 | |
| XM_004861472.2 | XP_004861529.1 | 3 - 4 | 1032 | 64 | ||
More Information
Set Membership: |
|

Rnf128
Genomic Map