Assay Details
| Gene Symbol: Tmem14c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847700.3 | XP_004847757.1 | 4 - 5 | 392 | 96 | |
| XM_004847701.2 | XP_004847758.1 | 4 - 5 | 410 | 96 | ||
More Information
Set Membership: |
|

Tmem14c
Genomic Map