Assay Details
| Gene Symbol: Pja2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841764.3 | XP_004841821.1 | 7 - 8 | 2003 | 67 | |
| XM_004841765.3 | XP_004841822.1 | 6 - 7 | 1817 | 67 | ||
More Information
Set Membership: |
|

Pja2
Genomic Map