Assay Details
| Gene Symbol: Cpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844206.3 | XP_004844263.1 | 5 - 6 | 608 | 78 | |
| XM_004844207.3 | XP_004844264.1 | 5 - 6 | 608 | 78 | ||
More Information
Set Membership: |
|

Cpb2
Genomic Map