Assay Details
| Gene Symbol: Aip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852231.3 | XP_004852288.1 | 2 - 3 | 514 | 62 | |
| XM_013075035.2 | XP_012930489.1 | 2 - 3 | 522 | 62 | ||
More Information
Set Membership: |
|

Aip
Genomic Map