Assay Details
| Gene Symbol: Alas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873550.3 | XP_004873607.1 | 10 - 11 | 1690 | 60 | |
| XM_004873552.1 | XP_004873609.1 | 9 - 10 | 1522 | 60 | ||
| XM_004873553.3 | XP_004873610.1 | 10 - 11 | 1598 | 60 | ||
| XM_004873554.3 | XP_004873611.1 | 9 - 10 | 1497 | 60 | ||
| XM_013068792.2 | XP_012924246.1 | 10 - 11 | 1583 | 60 | ||
| XM_013068794.1 | XP_012924248.1 | 9 - 10 | 1516 | 60 | ||
| XM_021251245.1 | XP_021106904.1 | 10 - 11 | 1685 | 60 | ||
| XM_021251246.1 | XP_021106905.1 | 10 - 11 | 1528 | 60 | ||
More Information
Set Membership: |
|

Alas2
Genomic Map