Assay Details
| Gene Symbol: Pitpna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857126.3 | XP_004857183.1 | 2 - 3 | 320 | 63 | |
| XM_021238347.1 | XP_021094006.1 | 2 - 3 | 203 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021238347.1 transcripts.
Set Membership: |
|

Pitpna
Genomic Map