Assay Details
| Gene Symbol: Fate1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871054.2 | XP_004871111.1 | 4 - 5 | 446 | 66 | |
| XM_021249158.1 | XP_021104817.1 | 4 - 5 | 623 | 66 | ||
| XM_021249159.1 | XP_021104818.1 | 4 - 5 | 540 | 66 | ||
| XM_021249160.1 | XP_021104819.1 | 4 - 5 | 542 | 66 | ||
More Information
Set Membership: |
|

Fate1
Genomic Map