Assay Details
| Gene Symbol: Setd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848508.2 | XP_004848565.1 | 1 - 2 | 153 | 70 | |
| XM_004848509.2 | XP_004848566.1 | 1 - 2 | 153 | 70 | ||
More Information
Set Membership: |
|

Setd9
Genomic Map