Assay Details
| Gene Symbol: Scfd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849721.3 | XP_004849778.1 | 2 - 3 | 1126 | 68 | |
| XM_021262346.1 | XP_021118005.1 | 2 - 3 | 1126 | 68 | ||
| XM_021262347.1 | XP_021118006.1 | 2 - 3 | 1126 | 68 | ||
| XM_021262348.1 | XP_021118007.1 | 2 - 3 | 1126 | 68 | ||
More Information
Set Membership: |
|

Scfd2
Genomic Map