Assay Details
| Gene Symbol: Slc38a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256651.1 | XP_021112310.1 | 2 - 3 | 572 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc38a8| Gene Symbol: Slc38a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256651.1 | XP_021112310.1 | 2 - 3 | 572 | 56 | |
Set Membership: |
|