Assay Details
| Gene Symbol: Stk16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864544.3 | XP_004864601.1 | 7 - 8 | 1071 | 62 | |
| XM_004864545.3 | XP_004864602.1 | 8 - 9 | 1336 | 62 | ||
| XM_004864546.3 | XP_004864603.1 | 7 - 8 | 993 | 62 | ||
| XM_021244209.1 | XP_021099868.1 | 7 - 8 | 891 | 62 | ||
| XM_021244210.1 | XP_021099869.1 | 5 - 6 | 640 | 62 | ||
More Information
Set Membership: |
|

Stk16
Genomic Map