Assay Details
| Gene Symbol: Snapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865385.3 | XP_004865442.1 | 2 - 3 | 1132 | 65 | |
| XM_004865387.2 | XP_004865444.1 | 2 - 3 | 176 | 65 | ||
More Information
Set Membership: |
|

Snapc2
Genomic Map