Assay Details
| Gene Symbol: Snapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865385.3 | XP_004865442.1 | 4 - 5 | 1624 | 75 | |
| XM_004865387.2 | XP_004865444.1 | 4 - 5 | 668 | 75 | ||
More Information
Set Membership: |
|

Snapc2
Genomic Map