Assay Details
| Gene Symbol: Fam19a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021266018.1 | XP_021121677.1 | 3 - 4 | 351 | 84 | |
| XM_021266019.1 | XP_021121678.1 | 3 - 4 | 567 | 84 | ||
| XM_021266020.1 | XP_021121679.1 | 2 - 3 | 434 | 84 | ||
| XM_021266021.1 | XP_021121680.1 | 3 - 4 | 351 | 84 | ||
More Information
Set Membership: |
|

Fam19a4
Genomic Map