Assay Details
| Gene Symbol: Eif2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013071837.2 | XP_012927291.1 | 6 - 7 | 649 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif2b1| Gene Symbol: Eif2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013071837.2 | XP_012927291.1 | 6 - 7 | 649 | 62 | |
Set Membership: |
|