Assay Details
| Gene Symbol: Prickle1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863171.3 | XP_004863228.1 | 5 - 6 | 848 | 75 | |
| XM_004863172.3 | XP_004863229.1 | 5 - 6 | 851 | 75 | ||
| XM_013079070.2 | XP_012934524.1 | 5 - 6 | 799 | 75 | ||
| XM_013079071.2 | XP_012934525.1 | 5 - 6 | 796 | 75 | ||
| XM_021243008.1 | XP_021098667.1 | 5 - 6 | 884 | 75 | ||
| XM_021243009.1 | XP_021098668.1 | 5 - 6 | 659 | 75 | ||
| XM_021243010.1 | XP_021098669.1 | 5 - 6 | 768 | 75 | ||
| XM_021243011.1 | XP_021098670.1 | 5 - 6 | 658 | 75 | ||
More Information
Set Membership: |
|

Prickle1
Genomic Map