Assay Details
| Gene Symbol: Cdkn2aip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852982.3 | XP_004853039.1 | 1 - 2 | 542 | 85 | |
| XM_021264971.1 | XP_021120630.1 | 1 - 2 | 561 | 85 | ||
More Information
Set Membership: |
|

Cdkn2aip
Genomic Map