Assay Details
| Gene Symbol: Fam19a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861139.3 | XP_004861196.1 | 2 - 3 | 1579 | 66 | |
| XM_004861140.3 | XP_004861197.1 | 3 - 4 | 1680 | 66 | ||
| XM_004861141.3 | XP_004861198.1 | 3 - 4 | 387 | 66 | ||
| XM_021241353.1 | XP_021097012.1 | 4 - 5 | 487 | 66 | ||
More Information
Set Membership: |
|

Fam19a2
Genomic Map