Assay Details
| Gene Symbol: Bud23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840052.3 | XP_004840109.1 | 10 - 11 | 784 | 52 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bud23| Gene Symbol: Bud23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840052.3 | XP_004840109.1 | 10 - 11 | 784 | 52 | |
Set Membership: |
|