Assay Details
| Gene Symbol: Dhx32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838741.3 | XP_004838798.1 | 10 - 11 | 2356 | 60 | |
| XM_004838742.3 | XP_004838799.1 | 10 - 11 | 2254 | 60 | ||
| XM_021253286.1 | XP_021108945.1 | 10 - 11 | 2253 | 60 | ||
| XM_021253287.1 | XP_021108946.1 | 9 - 10 | 1829 | 60 | ||
More Information
Set Membership: |
|

Dhx32
Genomic Map