Assay Details
Gene Symbol: Jazf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004839497.2 | XP_004839554.1 | 4 - 5 | 667 | 87 | |
XM_004839498.2 | XP_004839555.1 | 4 - 5 | 533 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
