Assay Details
| Gene Symbol: Ckmt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861701.3 | XP_004861758.1 | 3 - 4 | 289 | 80 | |
| XM_021241710.1 | XP_021097369.1 | 3 - 4 | 292 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004861701.3, XM_021241710.1 transcripts.
Set Membership: |
|

Ckmt1a
Genomic Map