Assay Details
| Gene Symbol: Ap3m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846316.2 | XP_004846373.1 | 5 - 6 | 1015 | 66 | |
| XM_021259640.1 | XP_021115299.1 | 5 - 6 | 1153 | 66 | ||
| XM_021259641.1 | XP_021115300.1 | 5 - 6 | 697 | 66 | ||
More Information
Set Membership: |
|

Ap3m1
Genomic Map