Assay Details
| Gene Symbol: Rcan3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850595.2 | XP_004850652.1 | 4 - 5 | 718 | 51 | |
| XM_021263072.1 | XP_021118731.1 | 4 - 5 | 620 | 51 | ||
More Information
Set Membership: |
|

Rcan3
Genomic Map