Assay Details
Gene Symbol: Tmem139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004860494.3 | XP_004860551.1 | 2 - 3 | 716 | 100 | |
XM_004860495.3 | XP_004860552.1 | 2 - 3 | 697 | 100 | ||
XM_013078013.2 | XP_012933467.1 | 3 - 4 | 519 | 100 | ||
XM_013078014.2 | XP_012933468.1 | 3 - 4 | 506 | 100 | ||
XM_021240943.1 | XP_021096602.1 | 2 - 3 | 433 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
