Assay Details
| Gene Symbol: Rpl36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865677.3 | XP_004865734.1 | 2 - 3 | 283 | 82 | |
| XM_004865678.3 | XP_004865735.1 | 2 - 3 | 215 | 82 | ||
More Information
Set Membership: |
|

Rpl36
Genomic Map