Assay Details
| Gene Symbol: Alg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854715.3 | XP_004854772.1 | 2 - 3 | 332 | 67 | |
| XM_004854716.3 | XP_004854773.1 | 2 - 3 | 407 | 67 | ||
| XM_004854718.3 | XP_004854775.1 | 2 - 3 | 332 | 67 | ||
More Information
Set Membership: |
|

Alg5
Genomic Map