Assay Details
| Gene Symbol: Dnal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845592.2 | XP_004845649.1 | 3 - 4 | 399 | 96 | |
| XM_004845593.3 | XP_004845650.1 | 3 - 4 | 393 | 96 | ||
More Information
Set Membership: |
|

Dnal4
Genomic Map