Assay Details
| Gene Symbol: Il1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844529.2 | XP_004844586.1 | 4 - 5 | 403 | 62 | |
| XM_021258166.1 | XP_021113825.1 | 4 - 5 | 445 | 62 | ||
More Information
Set Membership: |
|

Il1a
Genomic Map