Assay Details
| Gene Symbol: Guk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874725.3 | XP_004874782.1 | 5 - 6 | 500 | 88 | |
| XM_021252100.1 | XP_021107759.1 | 6 - 7 | 546 | 88 | ||
| XM_021252101.1 | XP_021107760.1 | 6 - 7 | 463 | 88 | ||
| XM_021252102.1 | XP_021107761.1 | 5 - 6 | 415 | 88 | ||
| XM_021252103.1 | XP_021107762.1 | 6 - 7 | 483 | 88 | ||
More Information
Set Membership: |
|

Guk1
Genomic Map