Assay Details
| Gene Symbol: Ly6e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837856.3 | XP_004837913.1 | 2 - 3 | 241 | 94 | |
| XM_021251244.1 | XP_021106903.1 | 3 - 4 | 278 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly6e| Gene Symbol: Ly6e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837856.3 | XP_004837913.1 | 2 - 3 | 241 | 94 | |
| XM_021251244.1 | XP_021106903.1 | 3 - 4 | 278 | 94 | ||