Assay Details
| Gene Symbol: Gabra4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849771.3 | XP_004849828.1 | 3 - 4 | 616 | 75 | |
| XM_004849773.2 | XP_004849830.1 | 2 - 3 | 145 | 75 | ||
| XM_021262393.1 | XP_021118052.1 | 3 - 4 | 629 | 75 | ||
More Information
Set Membership: |
|

Gabra4
Genomic Map