Assay Details
| Gene Symbol: Dph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871896.2 | XP_004871953.1 | 4 - 5 | 1365 | 67 | |
| XM_021249921.1 | XP_021105580.1 | 3 - 4 | 869 | 67 | ||
| XM_021249922.1 | XP_021105581.1 | 3 - 4 | 925 | 67 | ||
| XM_021249923.1 | XP_021105582.1 | 3 - 4 | 996 | 67 | ||
More Information
Set Membership: |
|

Dph2
Genomic Map