Assay Details
| Gene Symbol: Fam50a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875005.3 | XP_004875062.1 | 2 - 3 | 298 | 58 | |
| XM_021252332.1 | XP_021107991.1 | 2 - 3 | 296 | 58 | ||
More Information
Set Membership: |
|

Fam50a
Genomic Map