Assay Details
| Gene Symbol: Ssbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851302.3 | XP_004851359.2 | 3 - 4 | 296 | 62 | |
| XM_021263716.1 | XP_021119375.1 | 3 - 4 | 296 | 62 | ||
| XM_021263717.1 | XP_021119376.1 | 3 - 4 | 296 | 62 | ||
| XM_021263718.1 | XP_021119377.1 | 3 - 4 | 307 | 62 | ||
More Information
Set Membership: |
|

Ssbp1
Genomic Map